missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11303857
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-42-6
MDL Number MFCD00015996
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 863°C
Melting Point 567°C
Color White to Yellow
Physical Form Beads
Quantity 1 g
UN Number UN2570
Merck Index 14,1615
Formula Weight 272.22
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 5.192g/mL
Assay (metals basis)
Chemical Name or Material Cadmium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'