missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium carbonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 500°C (decomposition) |
Quantity | 250 g |
UN Number | UN2570 |
Merck Index | 14,1616 |
IUPAC Name | cadmium(2+);carbonate |
Formula Weight | 172.42 |
Percent Purity | 97% |
Odor | Odorless |
Density | 4.26 |
Chemical Name or Material | Cadmium carbonate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10244254 | Certificate of Analysis | 02/11/2022 | A15812.0B,A15812.30,A15812.18 |
10225597 | Certificate of Analysis | 23/11/2021 | A15812.0B,A15812.30,A15812.18 |
I2623C | Certificate of Analysis | 23/11/2021 | A15812.0B,A15812.30,A15812.18 |
J29S024 | Certificate of Analysis | 23/11/2021 | A15812.0B,A15812.30,A15812.18 |
A13R031A | Certificate of Analysis | 23/11/2021 | A15812.0B,A15812.30,A15812.18 |
Safety and Handling



Product Identifier
- Cadmium carbonate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H330-Fatal if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cadmium carbonate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cadmium carbonate, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction