Learn More
Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
60.90 € - 225.12 €
Chemical Identifiers
CAS | 513-78-0 |
---|---|
Molecular Formula | CCdO3 |
Molecular Weight (g/mol) | 172.42 |
MDL Number | MFCD00010918 |
InChI Key | GKDXQAKPHKQZSC-UHFFFAOYSA-L |
Synonym | cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3blyShow More |
PubChem CID | 10564 |
IUPAC Name | cadmium(2+);carbonate |
SMILES | [Cd++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360879
|
Thermo Scientific Alfa Aesar
010660.09 |
10 g |
60.90 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350879
|
Thermo Scientific Alfa Aesar
010660.18 |
50 g |
225.12 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadmium carbonate is the raw material for the production of pigments for ceramic and frits and for production of cadmium salt. Used as reagents. It is used for intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-78-0 | |
172.42 | |
GKDXQAKPHKQZSC-UHFFFAOYSA-L | |
10564 | |
[Cd++].[O-]C([O-])=O |
CCdO3 | |
MFCD00010918 | |
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3blyShow More | |
cadmium(2+);carbonate |
Specifications
500°C (decomposition) | |
White | |
99.998% (Metals basis) | |
MFCD00010918 | |
Soluble in dilute acids,concentrated solutions of ammonium salts. Insoluble in water. | |
[Cd++].[O-]C([O-])=O | |
172.42 | |
172.42 | |
4.26 g/cm3 |
513-78-0 | |
Powder | |
CCdO3 | |
cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly | |
GKDXQAKPHKQZSC-UHFFFAOYSA-L | |
cadmium(2+);carbonate | |
10564 | |
Odorless | |
Cadmium carbonate |
Safety and Handling
GHS H Statement
H302-H312-H332
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
H302+H312-H330-H340-H350-H361-H372
missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 208-168-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.