Learn More
Cadmium chloride, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10108-64-2 |
---|---|
Molecular Formula | CdCl2 |
Molecular Weight (g/mol) | 183.31 |
MDL Number | MFCD00010916 |
InChI Key | YKYOUMDCQGMQQO-UHFFFAOYSA-L |
Synonym | cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride |
PubChem CID | 24947 |
ChEBI | CHEBI:35456 |
SMILES | [Cl-].[Cl-].[Cd++] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10108-64-2 | |
183.31 | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
24947 | |
[Cl-].[Cl-].[Cd++] |
CdCl2 | |
MFCD00010916 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
CHEBI:35456 |
Specifications
960.0°C | |
10108-64-2 | |
Powder | |
CdCl2 | |
MFCD00010916 | |
15, 1622 | |
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether | |
[Cl-].[Cl-].[Cd++] | |
183.31 | |
CHEBI:35456 | |
99% | |
Cadmium chloride, 99% |
568.0°C | |
White | |
98.5% min. (On dry substance) (Argentometry) | |
CdCl2 | |
04,485 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
dichlorocadmium | |
24947 | |
183.31 | |
Analytical |
Safety and Handling
GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-296-7
missing translation for 'rtecsNumber' : EV0175000
missing translation for 'tsca' : TSCA
RUO – Research Use Only