missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ACS reagent, anhydrous, Thermo Scientific Chemicals

29.25 € - 347.73 €

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10682145
View Documents Promotion Details
Thermo Scientific Acros
315270050
5 g Glass bottle
29.25 €
5g
Please to purchase this item. Need a web account? Register with us today!
10412271
View Documents Promotion Details
Thermo Scientific Acros
315271000
100 g Plastic bottle
118.59 €
100g
Please to purchase this item. Need a web account? Register with us today!
10269843
View Documents Promotion Details
Thermo Scientific Acros
315275000
500 g Plastic bottle
347.73 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10108-64-2
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
CHEBI:35456
[Cl-].[Cl-].[Cd++]

Specifications

960.0°C
10108-64-2
Powder
CdCl2
04,485
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.31
ACS Reagent
568.0°C
White
CdCl2
MFCD00010916
15,1622
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in methanol and ethanol, practically insoluble in ether
[Cl-].[Cl-].[Cd++]
183.31
CHEBI:35456
≥99.0%
Cadmium chloride, Anhydrous, For analysis ACS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
May cause cancer.
May damage fertility.
May damage the unborn child.
Toxic if swallowed

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF S

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-296-7

Product Identifier
  • Cadmium chloride, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : cadmium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only