Learn More
Cadmium chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
123.00 € - 360.00 €
Chemical Identifiers
CAS | 10108-64-2 |
---|---|
Molecular Formula | CdCl2 |
Molecular Weight (g/mol) | 183.31 |
MDL Number | MFCD00010916 |
InChI Key | YKYOUMDCQGMQQO-UHFFFAOYSA-L |
Synonym | cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride |
PubChem CID | 24947 |
ChEBI | CHEBI:35456 |
IUPAC Name | dichlorocadmium |
SMILES | [Cl-].[Cl-].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301938
|
Thermo Scientific Alfa Aesar
011860.14 |
25 g |
123.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391928
|
Thermo Scientific Alfa Aesar
011860.22 |
100 g |
360.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadmium chloride is a precursor material for cadmium sulfide, which is a brilliant yellow pigment known as Cadmium Yellow. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium. It is a mild Lewis acid, and can form tetrachlorocadmium species. It is used in the preparation of organocadmium compounds, such as dialkyl or diaryl cadmium, which are useful for the synthesis of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10108-64-2 | |
183.31 | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
24947 | |
dichlorocadmium |
CdCl2 | |
MFCD00010916 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
CHEBI:35456 | |
[Cl-].[Cl-].[Cd++] |
Specifications
960°C | |
10108-64-2 | |
Powder | |
CdCl2 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
dichlorocadmium | |
24947 | |
183.32 | |
Hygroscopic | |
Cadmium chloride |
568°C | |
White | |
99.99% (Metals basis) | |
MFCD00010916 | |
Highly soluble in water. Soluble in dimethyl sulfoxide. Slightly soluble in alcohol. | |
[Cl-].[Cl-].[Cd++] | |
183.31 | |
CHEBI:35456 | |
Odorless | |
4.047 g/cm3 |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H335-H340-H350-H360FD-H372
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-296-7
missing translation for 'rtecsNumber' : EV0175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only