Learn More
Cadmium chloride, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals
108.54 € - 353.76 €
Chemical Identifiers
CAS | 10108-64-2 |
---|---|
Molecular Formula | CdCl2 |
Molecular Weight (g/mol) | 183.31 |
MDL Number | MFCD00010916 |
InChI Key | YKYOUMDCQGMQQO-UHFFFAOYSA-L |
Synonym | cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride |
PubChem CID | 24947 |
ChEBI | CHEBI:35456 |
IUPAC Name | dichlorocadmium |
SMILES | [Cl-].[Cl-].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338068
|
Thermo Scientific Alfa Aesar
036629.22 |
100 g |
108.54 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348068
|
Thermo Scientific Alfa Aesar
036629.36 |
500 g |
353.76 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadmium chloride is a precursor material for the preparation of cadmium sulfide. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Preparation of cadmium sulfide, as analytical lab reagent, photography, manufacturing of special mirrors, ice-nucleating agent, lubricant, to absorb H2S in sulfide analysis
Chemical Identifiers
10108-64-2 | |
183.31 | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
24947 | |
dichlorocadmium |
CdCl2 | |
MFCD00010916 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chlorideShow More | |
CHEBI:35456 | |
[Cl-].[Cl-].[Cd++] |
Specifications
960°C | |
10108-64-2 | |
CdCl2 | |
UN2570 | |
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride | |
YKYOUMDCQGMQQO-UHFFFAOYSA-L | |
dichlorocadmium | |
24947 | |
183.32 | |
ACS | |
Hygroscopic | |
Cadmium chloride, Anhydrous |
568°C | |
Powder | |
MFCD00010916 | |
14,1617 | |
Soluble in water,acetone; Slightly soluble in methanol; Insoluble in ether. | |
[Cl-].[Cl-].[Cd++] | |
183.31 | |
CHEBI:35456 | |
≤99.0% | |
Odorless | |
4.047 g/mL |
Safety and Handling
GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
H301-H330-H335-H340-H350-H360FD-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
missing translation for 'einecsNumber' : 233-296-7
missing translation for 'rtecsNumber' : EV0175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.