missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%, Thermo Scientific Chemicals

Product Code. 11391759
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cadmium chloride is also used for photocopying, dyeing and electroplating. Useful in pyrotechnic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-78-5
MDL Number MFCD00799071

Specifications

Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 79.5-81.0%
Molecular Formula CdCl2·2.5H2O
Solubility Information Highly soluble in water and slightly soluble in alcohol.
Formula Weight 228.36 (183.32 Anhydrous)
Grade ACS
Odor Odorless
Refractive Index 1.6513
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Cadmium chloride hemipentahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.