missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hemipentahydrate, ACS reagent

Product Code. p-45965 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10248563

Brand: Thermo Scientific Acros 423501000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-78-5
Molecular Formula 2·5 H2O
Molecular Weight (g/mol) 228.34
MDL Number MFCD00799071

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystals and/or Chunks
Quantity 100 g
Linear Formula CdCl2·5/2H2O
Merck Index 15, 1622
Solubility Information Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol
Formula Weight 228.34
Percent Purity 79.5 to 81.0% (CdCl2)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium chloride hemipentahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.