missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride, Honeywell™

Product Code. 15611880
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., anhydrous,≥99.0%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Anhydrous
Quantity 25 g
UN Number 2570
Solubility Information Soluble in water 457g/L at 20°C
Formula Weight 183.32g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Vapor Pressure 10mmHg (656°C)
Density Vapor Density: 6.3 (vs air)
Chemical Name or Material Cadmium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'