missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

64.62 € - 204.00 €

Chemical Identifiers

CAS 654054-66-7
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00149627
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
PubChem CID 16211517
IUPAC Name dichlorocadmium;hydrate
SMILES [Cl-].[Cl-].[Cd++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343597
View Documents
Thermo Scientific Alfa Aesar
020129.14
25 g
64.62 €
25g
Please to purchase this item. Need a web account? Register with us today!
11333597
View Documents
Thermo Scientific Alfa Aesar
020129.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Cadmium chloride hydrate is used in laboratory chemicals and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

654054-66-7
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
16211517
[Cl-].[Cl-].[Cd++]
CdCl2
MFCD00149627
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
dichlorocadmium;hydrate

Specifications

654054-66-7
CdCl2
UN2570
cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium;hydrate
16211517
99.99%
Hygroscopic
Cadmium chloride hydrate
Powder
MFCD00149627
14,1617
Soluble in water.
[Cl-].[Cl-].[Cd++]
183.31
183.32 (Anhydrous)
1.6513
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H340-H350-H360-H372
Fatal if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H340-H350-H360-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS

missing translation for 'einecsNumber' : 233-296-7

missing translation for 'rtecsNumber' : EV0175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only