missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

42.81 € - 1397.96 €

Chemical Identifiers

CAS 10108-64-2
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00010916
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
PubChem CID 24947
ChEBI CHEBI:35456
IUPAC Name dichlorocadmium
SMILES [Cl-].[Cl-].[Cd++]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11380078
SDS View Documents
Thermo Scientific Alfa Aesar
042852.04
2 g
42.81 €
2g
Please to purchase this item. Need a web account? Register with us today!
11310088
SDS View Documents
Thermo Scientific Alfa Aesar
042852.09
10 g
112.56 €
10g
Please to purchase this item. Need a web account? Register with us today!
11300088
SDS View Documents
Thermo Scientific Alfa Aesar
042852.18
50 g
361.80 €
50g
Please to purchase this item. Need a web account? Register with us today!
11390078
SDS View Documents
Thermo Scientific Alfa Aesar
042852.30
250 g
1397.96 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium chloride is used for the preparation of cadmium sulfide, used as Cadmium Yellow, a brilliant-yellow stable inorganic pigment. It can be used for the preparation of organocadmium compounds of the type R2Cd, where R is an aryl or a primary alkyl. It is also used for photocopying, dyeing and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10108-64-2
183.31
YKYOUMDCQGMQQO-UHFFFAOYSA-L
24947
dichlorocadmium
CdCl2
MFCD00010916
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
CHEBI:35456
[Cl-].[Cl-].[Cd++]

Specifications

960°C
10108-64-2
Powder
CdCl2
cadmium chloride, cadmium dichloride, caddy, vi-cad, kadmiumchlorid, caswell no. 135, ccris 114, hsdb 278, cadmium ii chloride
YKYOUMDCQGMQQO-UHFFFAOYSA-L
dichlorocadmium
24947
183.32
Hygroscopic
4.047 g/cm3 at 20°C
568°C
White
99.996% (Metal basis)
MFCD00010916
Soluble in water. Slightly soluble in alcohol.
[Cl-].[Cl-].[Cd++]
183.31
CHEBI:35456
Odorless
White Various forms (powder/flake/crystalline/beads
Cadmium chloride
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H335-H340-H350-H360FD-H372

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 233-296-7

missing translation for 'rtecsNumber' : EV0175000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.