missing translation for 'onlineSavingsMsg'
Learn More

Cadmium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

47.44 € - 376.88 €

Chemical Identifiers

CAS 7790-79-6
Molecular Formula CdF2
Molecular Weight (g/mol) 150.41
MDL Number MFCD00010919
InChI Key LVEULQCPJDDSLD-UHFFFAOYSA-L
IUPAC Name cadmium(2+) difluoride
SMILES [F-].[F-].[Cd++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349347
View Documents
Thermo Scientific Alfa Aesar
040203.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11369347
View Documents
Thermo Scientific Alfa Aesar
040203.14
25 g
137.69 €
25g
Please to purchase this item. Need a web account? Register with us today!
11359347
View Documents
Thermo Scientific Alfa Aesar
040203.22
100 g
376.88 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-79-6
150.41
LVEULQCPJDDSLD-UHFFFAOYSA-L
[F-].[F-].[Cd++]
CdF2
MFCD00010919
cadmium(2+) difluoride

Specifications

1758°C
7790-79-6
CdF2
UN2570
LVEULQCPJDDSLD-UHFFFAOYSA-L
cadmium(2+) difluoride
150.4
Puratronic™
6.64g/mL
Cadmium fluoride
1100°C
Powder
MFCD00010919
14,1619
[F-].[F-].[Cd++]
150.41
99.99%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.