missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 0.025mm (0.001 in.) thick, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11300649
Click to view available options
Quantity:
25 x 100 mm
25 x 400 mm
25 x 2000 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Quantity 25 x 2000 mm
Percent Purity 99.999%
Grade Puratronic™
Weight ≈0.54g/25x100mm
Form Foil
Assay (metals basis)
Chemical Name or Material Cadmium foil, 0.025mm (0.001 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'