missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis), Thermo Scientific Chemicals

64.82 € - 215.07 €

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
SMILES [Cd]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325839
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011370.FI
50 x 50 mm
64.82 €
Each
Please to purchase this item. Need a web account? Register with us today!
11391149
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011370.GH
100 x 100 mm
215.07 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-43-9
112.41
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
[Cd]
Cd
MFCD00010914
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
CHEBI:22977

Specifications

7440-43-9
MFCD00010914
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
99.9975%
≈5.4g/50x50mm
(metals basis)
Cd
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
[Cd]
112.41
CHEBI:22977
Puratronic™
Foil
Cadmium foil, 0.25mm (0.01 in.) thick
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c

H330-H341-H350-H361fd-H372-H500

missing translation for 'einecsNumber' : 231-152-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only