missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium hydroxide, 75%(as Cd), pure, Thermo Scientific™
Click to view available options
Quantity:
250 g
1 kg
Specifications
Specifications
Melting Point | 232.0°C |
Quantity | 250 g |
Assay Percent Range | 75%(as Cd) |
Merck Index | 15,1625 |
Solubility Information | Solubility in water: insoluble. Other solubilities: slightly sol in naoh sol, sol in dil acids, in nh4, or nh4cl sol |
Formula Weight | 146.41 |
Percent Purity | 75% (as Cd) |
Grade | Pure |
Chemical Name or Material | Cadmium hydroxide |
Safety and Handling
Product Identifier
- Cadmium hydroxide
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H330-Fatal if inhaled.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction