missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium hydroxide, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
25 g
Description
It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 300°C (decomposition) |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN2570 |
Merck Index | 14,1620 |
Solubility Information | Soluble in dilute acids. Slightly soluble in water; 0.026g/100 mL. |
Formula Weight | 146.41 |
Percent Purity | 99.98% |
Grade | Puratronic™ |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'