missing translation for 'onlineSavingsMsg'
Learn More

Cadmium hydroxide, Puratronic™, 99.98% (metals basis), Thermo Scientific Chemicals

Product Code. 11310889
Click to view available options
Quantity:
2 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 300°C (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN2570
Merck Index 14,1620
Solubility Information Soluble in dilute acids. Slightly soluble in water; 0.026g/100 mL.
Formula Weight 146.41
Percent Purity 99.98%
Grade Puratronic™
Sensitivity Air Sensitive
Density 4.79 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Cadmium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'