Learn More
Cadmium iodide, 99%, Thermo Scientific Chemicals
149.00 € - 461.00 €
Chemical Identifiers
CAS | 7790-80-9 |
---|---|
MDL Number | MFCD00010920 |
InChI Key | OKIIEJOIXGHUKX-UHFFFAOYSA-L |
Synonym | cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g |
PubChem CID | 24635 |
IUPAC Name | cadmium(2+);diiodide |
SMILES | [Cd+2].[I-].[I-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10481441
|
Thermo Scientific Acros
211462500 |
250 g | Glass bottle |
149.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10402855
|
Thermo Scientific Acros
211460010 |
1 kg | Glass bottle |
461.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-80-9 | |
OKIIEJOIXGHUKX-UHFFFAOYSA-L | |
24635 | |
[Cd+2].[I-].[I-] |
MFCD00010920 | |
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g | |
cadmium(2+);diiodide |
Specifications
7790-80-9 | |
Shiny Crystalline Flakes or Powder | |
CdI2 | |
MFCD00010920 | |
cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g | |
[Cd+2].[I-].[I-] | |
366.223 | |
366.2 | |
Cadmium iodide, 99% |
White to Yellow | |
99% | |
CdI2 | |
15, 1626 | |
OKIIEJOIXGHUKX-UHFFFAOYSA-L | |
cadmium(2+);diiodide | |
24635 | |
≥98.5% |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 232-223-6
missing translation for 'rtecsNumber' : EV1290000
missing translation for 'tsca' : TSCA
- Cadmium iodide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only