missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals

68.44 € - 217.08 €

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11351579
View Documents
Thermo Scientific Alfa Aesar
012228.22
100 g
68.44 €
100g
Please to purchase this item. Need a web account? Register with us today!
11346648
View Documents
Thermo Scientific Alfa Aesar
012228.36
500 g
217.08 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10022-68-1
238.44
IZIPRBSCGMISKX-UHFFFAOYSA-N
56924536
cadmium(2+);dinitrate;tetrahydrate
CdH2N2O6
MFCD00149626
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
CHEBI:86156
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

132°C
10022-68-1
Crystals
CdH2N2O6
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium(2+);dinitrate;tetrahydrate
56924536
308.47 (236.41 Anhydrous)
2.45
59°C
White
98.5% min.
MFCD00149626
Highly soluble in water. Soluble in alcohol,acetone,and ammonia.
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
238.44
CHEBI:86156
Weak
Cadmium nitrate tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H272-H312-H332

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H301-H312-H330-H340-H350-H360-H372

missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 233-710-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cadmium nitrate tetrahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Cadmium nitrate tetrahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.