missing translation for 'onlineSavingsMsg'
Learn More
Cadmium oxide, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11396628

Cadmium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11396628
500 g
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11396628

Brand: Thermo Scientific Alfa Aesar 012219.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium oxide is used in ceramic glazes, cadmium electroplating baths, pigments, phosphors, electrodes for storage batteries, cadmium salts, and heterogeneous catalysis for dehydrogenation. It is a basic conducting material used to prepare transparent conducting films, which finds use in phototransistors, liquid crystal displays, photodiodes, photovoltaic cells, and antireflection coatings. Electrodeposited cadmium oxide films have excellent electrocatalytic activity towards many organic compounds, ozone and oxygen evolution. CdO is selective in phenol photodegradation. Glassy carbon electrode coated with cadmium oxide film has been used to develop a sensitive and rapid assay method for penicillamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
PubChem CID 14782
SMILES [O--].[Cd++]

Specifications

Melting Point 1430°C
Color Red to Brown
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in dilute acid. Slowly soluble in ammonium salts. Insoluble in water.
IUPAC Name cadmium(2+) oxidandiide
Formula Weight 128.4
Odor Odorless
Density 8.15 g/cm3
Chemical Name or Material Cadmium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR04G019 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog Number012219.A3,012219.36
Lot NumberY05F020 Certificate TypeCertificate of Analysis Date01/06/2022 Catalog Number012219.A3,012219.36
Lot NumberQ27I039 Certificate TypeCertificate of Analysis Date03/05/2022 Catalog Number012219.A3,012219.36
Lot NumberM08H075 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012219.A3,012219.36
Lot NumberG07F26 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012219.A3,012219.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium oxide, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.