Learn More
Cadmium oxide, 99%, pure, Thermo Scientific Chemicals
39.09 € - 105.53 €
Chemical Identifiers
CAS | 1306-19-0 |
---|---|
Molecular Formula | CdO |
Molecular Weight (g/mol) | 128.41 |
MDL Number | MFCD00010921 |
InChI Key | CFEAAQFZALKQPA-UHFFFAOYSA-N |
Synonym | cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 |
PubChem CID | 14782 |
IUPAC Name | oxocadmium |
SMILES | [O--].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15299716
|
Thermo Scientific Acros
223792500 |
250 g |
39.09 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15279716
|
Thermo Scientific Acros
223790010 |
1 kg |
105.53 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-19-0 | |
128.41 | |
CFEAAQFZALKQPA-UHFFFAOYSA-N | |
14782 | |
[O--].[Cd++] |
CdO | |
MFCD00010921 | |
cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 | |
oxocadmium |
Specifications
1385.0°C | |
98.5% min. (Complexometry) | |
CdO | |
15,1628 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted acids and alkalies | |
[O--].[Cd++] | |
128.41 | |
128.41 | |
Pure |
1306-19-0 | |
CdO | |
MFCD00010921 | |
cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 | |
CFEAAQFZALKQPA-UHFFFAOYSA-N | |
oxocadmium | |
14782 | |
99% | |
Cadmium oxide |
Safety and Handling
GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
May cause cancer.
Suspected
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-146-2
- Cadmium oxide, brown
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H330-Fatal if inhaled.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.