Learn More
Cadmium oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Cadmium oxide is used in ceramic glazes, cadmium electroplating baths, pigments, phosphors, electrodes for storage batteries, cadmium salts, and heterogeneous catalysis for dehydrogenation. It is a basic conducting material used to prepare transparent conducting films, which finds use in phototransistors, liquid crystal displays, photodiodes, photovoltaic cells, and antireflection coatings. Electrodeposited cadmium oxide films have excellent electrocatalytic activity towards many organic compounds, ozone and oxygen evolution. CdO is selective in phenol photodegradation. Glassy carbon electrode coated with cadmium oxide film has been used to develop a sensitive and rapid assay method for penicillamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1430°C |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in dilute acid. Slowly soluble in ammonium salts. Insoluble in water. |
Formula Weight | 128.4 |
Density | 8.15 |
Chemical Name or Material | Cadmium oxide |
Safety and Handling
- Cadmium oxide, Puratronic
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cadmium oxide
RUO – Research Use Only