missing translation for 'onlineSavingsMsg'
Learn More

Cadmium phosphate (ortho), 99%, Thermo Scientific Chemicals

Product Code. 11319347
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13847-17-1
Molecular Formula Cd3O8P2
Molecular Weight (g/mol) 527.182
MDL Number MFCD00049414
InChI Key NRGIRRZWCDKDMV-UHFFFAOYSA-H
Synonym cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate
PubChem CID 26060
IUPAC Name cadmium(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2]

Specifications

Melting Point ∼1500°C
Physical Form Powder
Quantity 10 g
UN Number UN2570
Solubility Information Insoluble in water
Formula Weight 527.14
Percent Purity 99%
Chemical Name or Material Cadmium phosphate (ortho)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'