missing translation for 'onlineSavingsMsg'
Learn More

Cadmium powder, -200 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

90.95 € - 775.86 €

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
SMILES [Cd]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331129
View Documents
Thermo Scientific Alfa Aesar
011353.06
5 g
90.95 €
5g
Please to purchase this item. Need a web account? Register with us today!
11351129
View Documents
Thermo Scientific Alfa Aesar
011353.14
25 g
232.16 €
25g
Please to purchase this item. Need a web account? Register with us today!
11341129
View Documents
Thermo Scientific Alfa Aesar
011353.22
100 g
775.86 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-43-9
112.41
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
23973
[Cd]
Cd
MFCD00010914
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
CHEBI:22977

Specifications

White
Cd
UN3179
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium(2+) ion
23973
99.99%
Solid
Cadmium powder
7440-43-9
MFCD00010914
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
[Cd]
112.41
CHEBI:22977
−200 Mesh
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.

P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c

H228-H330-H341-H350-H361fd-H372

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-152-8

missing translation for 'rtecsNumber' : EU9800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.