Learn More
Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7790-84-3 |
---|---|
Molecular Formula | Cd3O12S3·8H2O |
Molecular Weight (g/mol) | 769.51 |
MDL Number | MFCD02253000 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10094063
|
Thermo Scientific Acros
403761000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
10616431
|
Thermo Scientific Acros
403765000 |
500 g | Glass bottle |
N/A
|
N/A | ||||
11944241
|
Thermo Scientific Acros
403760025 |
2.5 kg | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7790-84-3 | |
Cd3O12S3·8H2O | |
769.51 | |
MFCD02253000 |
Specifications
41.0°C | |
White | |
Cd3O12S3·8H2O | |
MFCD02253000 | |
Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol,acids,acetone and ammonia | |
769.51 | |
ACS Reagent |
7790-84-3 | |
Crystals | |
3CdSO4·8H2O | |
15,1632 | |
769.51 | |
98.0 to 102.0% | |
Cadmium sulfate, 8/3-hydrate |
Safety and Handling
GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust
GHS Signal Word: Danger
RUO – Research Use Only