missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate 8/3-Hydrate, puriss. p.a. ACS Reagent, ≥99.0%, Honeywell Fluka™

Product Code. 15691890
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (calc. based on CdSO4 · 8/3H2O, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD02253000

Specifications

Melting Point 41.5°C
Color Colorless
Physical Form Solid
Quantity 500 g
Linear Formula CdSO4 · 8/3H2O
UN Number UN2570
Formula Weight 256.57g/mol
Percent Purity ≥99.0% (calc. based on CdSO4.83H2O, KT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Density 3.09g/cu.cm
Chemical Name or Material Cadmium Sulfate 8/3-Hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.