missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate, anhydrous, ACS, 99+%, Thermo Scientific Chemicals

29.25 € - 86.03 €

Chemical Identifiers

CAS 10124-36-4
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00010923
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
Synonym cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
PubChem CID 24962
ChEBI CHEBI:50292
IUPAC Name cadmium(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Cd+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321938
SDS View Documents
Thermo Scientific Alfa Aesar
011861.14
25 g
29.25 €
25g
Please to purchase this item. Need a web account? Register with us today!
11311938
SDS View Documents
Thermo Scientific Alfa Aesar
011861.22
100 g
86.03 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium sulfate is used in pigments and electroplating. It acts as an accelerator in cement formation. It is also employed as lubricants. Further, it is useful for the electroplating of cadmium in electronic circuits. It serves as a precursor to prepare cadmium-based pigment like cadmium sulfide. In the Western standard cell, it is used for electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10124-36-4
208.47
QCUOBSQYDGUHHT-UHFFFAOYSA-L
24962
cadmium(2+);sulfate
CdO4S
MFCD00010923
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
CHEBI:50292
[O-]S(=O)(=O)[O-].[Cd+2]

Specifications

1000°C
White
>99%
MFCD00010923
Soluble in water. Slightly soluble in methanol and ethyl acetate. Insoluble in ethanol.
[O-]S(=O)(=O)[O-].[Cd+2]
208.47
CHEBI:50292
ACS
White powder
Cadmium sulfate, For analysis ACS
10124-36-4
Powder
CdO4S
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium(2+);sulfate
24962
208.46
Hygroscopic
4.691
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P280-P301+P310-P304+P340-P308+P313-P310-P330-P501c

H301-H330-H340-H350-H360FD-H372

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III

missing translation for 'einecsNumber' : 233-331-6

missing translation for 'rtecsNumber' : EV2700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.