missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Sulfate, Extra Pure, SLR, Fisher Chemical™
71.60 € - 241.00 €
Chemical Identifiers
CAS | 7790-84-3 |
---|---|
Molecular Formula | CdH2O5S |
Molecular Weight (g/mol) | 226.49 |
MDL Number | 2253000 |
InChI Key | VRFRVKRTIMIRNG-UHFFFAOYSA-L |
IUPAC Name | cadmium(2+) hydrate sulfate |
SMILES | O.[Cd++].[O-]S([O-])(=O)=O |
Description
Chemical Identifiers
7790-84-3 | |
226.49 | |
VRFRVKRTIMIRNG-UHFFFAOYSA-L | |
O.[Cd++].[O-]S([O-])(=O)=O |
CdH2O5S | |
2253000 | |
cadmium(2+) hydrate sulfate |
Specifications
41°C | |
White | |
Solid | |
2253000 | |
O.[Cd++].[O-]S([O-])(=O)=O | |
226.49 | |
Plastic powder jar |
7790-84-3 | |
4.5 to 6.0 | |
CdH2O5S | |
VRFRVKRTIMIRNG-UHFFFAOYSA-L | |
cadmium(2+) hydrate sulfate | |
769.51g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cadmium sulfate octahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 2
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H350-May cause cancer.
- H340-May cause genetic defects.
- H360FD-May damage fertility. May damage the unborn child.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title