missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11479452
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfate hydrate is used for formulating screens or for optimization. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water (1130g/L ).

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-35-6
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00151655
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

Melting Point 41.5°C (trans pt)
Quantity 100 g
UN Number UN2570
Merck Index 14,1627
Solubility Information Soluble in water (1130g/L ).
Formula Weight 208.46 (Anhydrous)
Percent Purity 98%
Odor Odorless
Refractive Index 1.565
Density 3.08
Chemical Name or Material Cadmium sulfate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'