Learn More
Cadmium sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Vacuum deposition
392.96 € - 1199.97 €
Chemical Identifiers
CAS | 1306-23-6 |
---|---|
Molecular Formula | CdS |
Molecular Weight (g/mol) | 144.47 |
MDL Number | MFCD00010922 |
InChI Key | FRLJSGOEGLARCA-UHFFFAOYSA-N |
Synonym | cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow |
PubChem CID | 14783 |
ChEBI | CHEBI:50833 |
IUPAC Name | sulfanylidenecadmium |
SMILES | [S--].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344258
|
Thermo Scientific Alfa Aesar
041511.14 |
25 g |
392.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334258
|
Thermo Scientific Alfa Aesar
041511.22 |
100 g |
1199.97 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vacuum deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-23-6 | |
144.47 | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
14783 | |
sulfanylidenecadmium |
CdS | |
MFCD00010922 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
CHEBI:50833 | |
[S--].[Cd++] |
Specifications
980°C (sublimation) | |
Orange | |
CdS | |
UN3077 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
[S--].[Cd++] | |
144.47 | |
CHEBI:50833 | |
99.99% | |
4.82 g/mL | |
Cadmium sulfide |
1306-23-6 | |
Sintered Lump | |
MFCD00010922 | |
14,1628 | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
sulfanylidenecadmium | |
14783 | |
144.46 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H350-H361fd-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-147-8
missing translation for 'rtecsNumber' : EV3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only