missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15452267
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum.

Cadmium sulfide is used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. It is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
IUPAC Name sulfanylidenecadmium
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Physical Form Pieces
Quantity 50 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 144.46
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 4.82
Density 4.82
Chemical Name or Material Cadmium sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.