missing translation for 'onlineSavingsMsg'
Learn More

Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Semiconductors, phosphors

28.40 € - 183.92 €

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307927
View Documents
Thermo Scientific Alfa Aesar
014367.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11397917
View Documents
Thermo Scientific Alfa Aesar
014367.06
5 g
61.61 €
5g
Please to purchase this item. Need a web account? Register with us today!
11387917
View Documents
Thermo Scientific Alfa Aesar
014367.14
25 g
183.92 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Semiconductors, phosphorsCadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1306-25-8
240.01
RPPBZEBXAAZZJH-UHFFFAOYSA-N
91501
[Cd]=[Te]
CdTe
MFCD00015998
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
tellanylidenecadmium

Specifications

1041°C
Black
CdTe
UN3077
cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
RPPBZEBXAAZZJH-UHFFFAOYSA-N
tellanylidenecadmium
91501
99.999%
6.20 g/mL
Cadmium telluride
1306-25-8
Polycrystalline Powder
MFCD00015998
14,1629
Insoluble in water and acids except HNO3 in which it is soluble with decomposition.
[Cd]=[Te]
240.01
240
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P501c

H332-H350-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-149-9

missing translation for 'rtecsNumber' : EV3330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only