Learn More
Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
143.00 € - 435.00 €
Chemical Identifiers
CAS | 7440-43-9 |
---|---|
Molecular Formula | Cd |
Molecular Weight (g/mol) | 112.41 |
MDL Number | MFCD00010914 |
InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
Synonym | kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds |
PubChem CID | 23973 |
ChEBI | CHEBI:22977 |
IUPAC Name | cadmium |
SMILES | [Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380639
|
Thermo Scientific Alfa Aesar
000060.G4 |
3 m |
143.00 €
3m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390639
|
Thermo Scientific Alfa Aesar
000060.G7 |
15 m |
435.00 €
15m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-43-9 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
23973 | |
cadmium |
Cd | |
MFCD00010914 | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
CHEBI:22977 | |
[Cd] |
Specifications
7440-43-9 | |
MFCD00010914 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
99.999% | |
≈2.8g/m | |
(metals basis) |
Cd | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
Puratronic™ | |
Wire | |
Cadmium wire, 0.64mm (0.025 in.) dia. |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
H330-H341-H350-H361fd-H372-H500
missing translation for 'einecsNumber' : 231-152-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cadmium wire
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P201-Obtain special instructions before use.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only