missing translation for 'onlineSavingsMsg'
Learn More

Caesium Chloride, 99.5+%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10213710
100 g
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ClCs, CAS Number-7647-17-8, 1kg, 1290 deg.C, White, 168.36g/mol, 646 deg.C, 10955, Plastic powder jar, 5 to 9, Powder Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290°C
Melting Point 646°C
Color White
pH 5 to 9
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 168.36g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Caesium Chloride, 99.5+%, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.