Learn More
Caffeine, 99.7%, Thermo Scientific Chemicals
Description
It acts as a stimulant drug and a reversible acetylcholinesterase inhibitor. It plays an important role for the inactivation of Escherichia coli O 157:H 7 in brain heart infusion (BHI) broth. It is also useful for the protection from photodamage after ultraviolet irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 234°C to 236°C |
Density | 1.23 g/mL |
Boiling Point | 178°C (sublimation) |
Assay Percent Range | 99.7% |
Quantity | 25 g |
Beilstein | 17705 |
Solubility Information | Soluble in water,pyridine and ethyl acetate. Partially soluble in acetone,alcohol and petroleum ether. |
Formula Weight | 194.19 |
Physical Form | Powder |
Chemical Name or Material | Caffeine |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
R06K031 | Certificate of Analysis | 03/03/2025 | 039214.14,039214.36,039214.22 |
W23K006 | Certificate of Analysis | 09/10/2024 | 039214.14,039214.36,039214.22 |
S03K029 | Certificate of Analysis | 11/06/2024 | 039214.14,039214.36,039214.22 |
X11J003 | Certificate of Analysis | 16/10/2023 | 039214.14,039214.36,039214.22 |
P03H045 | Certificate of Analysis | 29/08/2022 | 039214.14,039214.36,039214.22 |
Safety and Handling

- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.