Learn More
Caframo™ Impeller Shaft
GREENER_CHOICE
Stainless steel Impeller Shaft designed for use with Caframo™ Impeller and Propeller blades. Maximum corrosion protection available in multiple sizes.
71.35 € - 187.00 €
Specifications
Type Stirring Shaft
Material Stainless Steel (316L)
View More Specs
Products 9
Product Code Brand Length (Metric) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Price Quantity & Availability  
12907136
GREENER_CHOICE
View Documents
Caframo™
A712
305 mm 8 mm
71.35 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
12917136
View Documents
Caframo™
A713
305 mm 9.5 mm
87.30 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
12967106
GREENER_CHOICE
View Documents
Caframo™
A162
406 mm 8 mm
85.20 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
12927136
GREENER_CHOICE
View Documents
Caframo™
A722
457 mm 8 mm
102.00 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
12937136
View Documents
Caframo™
A723
457 mm 9.5 mm
102.00 €
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
12947136
GREENER_CHOICE
View Documents
Caframo™
A733
508 mm 9.5 mm
128.00 €
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
12957136
GREENER_CHOICE
View Documents
Caframo™
A742
610 mm 8 mm
136.00 €
Each
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
12967136
GREENER_CHOICE
View Documents
Caframo™
A743
610 mm 9.5 mm
154.00 €
Each
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
12977136
GREENER_CHOICE
View Documents
Caframo™
A753
762 mm 9.5 mm
187.00 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
  • Electro-polished stainless steel 316L
  • 9.5 mm Shafts have a turned down end to 8 mm
  • Compatible with all Caframo™ and Fisherbrand™ Overhead Stirrers
Specifications
Specifications
Stirring Shaft
Stainless Steel (316L)