Learn More
IKA™ CAL-O-1000 Silicon Oil
Certified standard silicon oil designed to allow you to check the measurement accuracy of your ROTAVISC.
Brand: IKA™ 0025000436
166.25 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes certificate
- For use with a range of ROTAVISC models
Specifications
Standard Silicon Oil | |
Period of Validity: 12 months | |
500 mL |
1000 cSt Dynamic | |
25°C |