Learn More
Calcifediol-d6, MedChemExpress
MedChemExpress Calcifediol-d6 is the deuterium labeled Calcifediol. Calcifediol, a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
Brand:  Medchem Express HY-13332
Encompass_Preferred
 View more versions of this product
Product Code. 16471339
  • 530.55 € / 1mg
Explore more special offers
Description
Description
Calcifediol-d6 is the deuterium labeled Calcifediol. Calcifediol, a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
Specifications
Specifications
Calcifediol-d6
1 mg
Research
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
406.67
Research
Solid
78782-98-6
White
C27H38D6O2
406.67
98.0%
Metabolism-sugar/lipid metabolism