Learn More
Calcitriol-d6, MedChemExpress
MedChemExpress Calcitriol-d6 is the deuterium labeled Calcitriol. Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).
Brand:  Medchem Express HY-76814
Encompass_Preferred
Product Code. 16431979
  • 996.30 € / 1mg
Explore more special offers
Description
Description
Calcitriol-d6 is the deuterium labeled Calcitriol. Calcitriol is the active metabolite of vitamin D3 that activates the vitamin D receptor (VDR).
Specifications
Specifications
Calcitriol-d6
1 mg
Research
DMSO : 50 mg/mL (118.30 mM; Need ultrasonic)
422.67
99.12%
Metabolism-sugar/lipid metabolism
78782-99-7
White
C27H38D6O3
C=C1/C(C[C@H](C[C@@H]1O)O)=C\C=C2CCC[C@]3(C)[C@@H]([C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)CC[C@@]/23[H]
422.67
Research
Solid