missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

60.70 € - 217.08 €

Chemical Identifiers

CAS 114460-21-8
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362068
View Documents
Thermo Scientific Alfa Aesar
012974.06
5 g
60.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11352068
View Documents
Thermo Scientific Alfa Aesar
012974.14
25 g
217.08 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium acetate has been utilized in the crystallization of proteins, such as human annexin II and the bacterial heme oxygenase (Hmu O), and of a complex of human CDK6 with a virus-encoded cyclin. It has been used in the manufactureof metallic soaps and synthetic resins, in lubricants as a corrosion inhibitor, and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

114460-21-8
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
[Ca++].CC([O-])=O.CC([O-])=O
C4H6CaO4
MFCD00012448
calcium diacetate

Specifications

(decomposition)
Powder
MFCD00012448
14,1646
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
158.17 (Anhydrous)
Puratronic™
1.5 g/mL
Calcium acetate hydrate
114460-21-8
C4H6CaO4
3692527
Soluble in water. Slightly soluble in methanol,hydrazine insoluble in acetone,ethanol and benzene.
[Ca++].CC([O-])=O.CC([O-])=O
158.17
99.9965%
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-540-9

missing translation for 'rtecsNumber' : AF7525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only