missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate Monohydrate, ACS Reagent, ≥99%, Honeywell Fluka™

Product Code. p-7120278 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15695400

Brand: Honeywell-Fluka 402850100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
MDL Number MFCD00150019
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Color White
Quantity 100 g
Linear Formula Ca(CH3CO2)2 · H2O
UN Number NONH for all modes of transport
Formula Weight 176.18g/mol
Percent Purity ≥99%
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Calcium acetate monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.