missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
54.80 € - 158.00 €
Chemical Identifiers
CAS | 5743-26-0 |
---|---|
Molecular Formula | C4H6CaO4·H2O |
Molecular Weight (g/mol) | 176.19 |
InChI Key | XQKKWWCELHKGKB-UHFFFAOYSA-L |
Synonym | calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu |
PubChem CID | 82163 |
ChEBI | CHEBI:59199 |
IUPAC Name | calcium;diacetate;hydrate |
SMILES | CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12625027
|
Thermo Scientific Acros
446351000 |
100 g | Plastic bottle |
54.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12645027
|
Thermo Scientific Acros
446355000 |
500 g | Plastic bottle |
158.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5743-26-0 | |
176.19 | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
CHEBI:59199 | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
C4H6CaO4·H2O | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
82163 | |
calcium;diacetate;hydrate |
Specifications
160.0°C | |
C4H6CaO4·H2O | |
15,1649 | |
Solubility in water: soluble | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] | |
176.19 | |
CHEBI:59199 | |
ACS Reagent |
5743-26-0 | |
(CH3CO2)2Ca·H2O | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
calcium;diacetate;hydrate | |
82163 | |
176.19 | |
Calcium acetate monohydrate |
RUO – Research Use Only