Learn More
Calcium arsenate, 95%, Thermo Scientific Chemicals
122.61 € - 470.34 €
Chemical Identifiers
CAS | 7778-44-1 |
---|---|
Molecular Formula | As2Ca3O8 |
Molecular Weight (g/mol) | 398.069 |
MDL Number | MFCD00049407 |
InChI Key | RMBBSOLAGVEUSI-UHFFFAOYSA-H |
Synonym | calcium arsenate, pencal, cucumber dust, spra-cal, calcium-o-arsenate, chip-cal granular, tricalcium arsenate, calcium orthoarsenate, kalziumarseniat german, arsenic acid, calcium salt |
PubChem CID | 24501 |
IUPAC Name | tricalcium;trioxido(oxo)-$l^{5}-arsane |
SMILES | [O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11323618
|
Thermo Scientific Alfa Aesar
039219.18 |
50 g | Packing group II |
122.61 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11313618
|
Thermo Scientific Alfa Aesar
039219.30 |
250 g | Packing group II |
470.34 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Calcium arsenate is also used in weather- resistant wood treatment. It was used mainly in viticulture, in the past. it was mainly used against the boll weevil in cotton cultivation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-44-1 | |
398.069 | |
RMBBSOLAGVEUSI-UHFFFAOYSA-H | |
24501 | |
[O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] |
As2Ca3O8 | |
MFCD00049407 | |
calcium arsenate, pencal, cucumber dust, spra-cal, calcium-o-arsenate, chip-cal granular, tricalcium arsenate, calcium orthoarsenate, kalziumarseniat german, arsenic acid, calcium salt | |
tricalcium;trioxido(oxo)-$l^{5}-arsane |
Specifications
7778-44-1 | |
Powder | |
As2Ca3O8 | |
calcium arsenate, pencal, cucumber dust, spra-cal, calcium-o-arsenate, chip-cal granular, tricalcium arsenate, calcium orthoarsenate, kalziumarseniat german, arsenic acid, calcium salt | |
RMBBSOLAGVEUSI-UHFFFAOYSA-H | |
tricalcium;trioxido(oxo)-$l^{5}-arsane | |
24501 | |
3.62 g/cm3 at 20°C |
White | |
95% | |
MFCD00049407 | |
It is soluble in water and acids,insoluble in organic solvents | |
[O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2] | |
398.069 | |
398.03 | |
Calcium arsenate |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-904-5
missing translation for 'rtecsNumber' : CG0830000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.