missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium benzoate hydrate, 98%, water <8%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Calcium benzoate hydrate is used in used in the food industry as a preservative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility
Soluble in water (2.6 g/100ml at 0°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Calcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility
Soluble in water (2.6 g/100ml at 0°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Molecular Formula | C14H10CaO4 |
MDL Number | MFCD09953675 |
Quantity | 500 g |
Synonym | calcium benzoate, calcium dibenzoate, unii-3qde968mkd, 3qde968mkd, benzoic acid, calcium salt, benzoic acid calcium, acmc-20alj1, calcium 2+ dibenzoate, calcium benzoate, reagent, e213 benzoic acid |
Solubility Information | Soluble in water (2.6g/100ml at 0°C). |
InChI Key | NSQPPSOSXWOZNH-UHFFFAOYSA-L |
SMILES | [Ca++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1 |
IUPAC Name | calcium;dibenzoate |
Molecular Weight (g/mol) | 282.31 |
PubChem CID | 62425 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction