missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, 95%, Thermo Scientific Chemicals

37.19 € - 231.15 €

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343487
View Documents
Thermo Scientific Alfa Aesar
018820.06
5 g
37.19 €
5g
Please to purchase this item. Need a web account? Register with us today!
11353487
View Documents
Thermo Scientific Alfa Aesar
018820.22
100 g
80.40 €
100g
Please to purchase this item. Need a web account? Register with us today!
11333487
View Documents
Thermo Scientific Alfa Aesar
018820.36
500 g
231.15 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium bromide hydrate is used as freezing mixtures, food preservatives and photography. Its aqueous solution is used in drilling fluids and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71626-99-8
199.89
WGEFECGEFUFIQW-UHFFFAOYSA-L
[Ca++].[Br-].[Br-]
Br2Ca
MFCD00149608
calcium dibromide

Specifications

71626-99-8
Powder
Br2Ca
Soluble in water.
[Ca++].[Br-].[Br-]
199.89
Odorless
Calcium bromide hydrate
White
95.0%
MFCD00149608
WGEFECGEFUFIQW-UHFFFAOYSA-L
calcium dibromide
199.90 (Anhydrous)
White powder
Videos
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

missing translation for 'einecsNumber' : 232-164-6

missing translation for 'rtecsNumber' : EV9328000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only