missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, extra pure, Thermo Scientific Chemicals

Product Code. 15109323
Click to view available options
Quantity:
250 g
2.5 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Melting Point 730.0°C
Quantity 2.5 kg
Linear Formula CaBr2·xH2O
Solubility Information (10% in water) Clear colorless to slightly hazy
Formula Weight 199.9
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Calcium bromide hydrate
compliance-icons
Product Identifier
  • Calcium bromide hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Calcium bromide, hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'