missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium bromide hydrate, extra pure, Thermo Scientific Chemicals
51.40 € - 292.00 €
Chemical Identifiers
CAS | 71626-99-8 |
---|---|
Molecular Formula | Br2Ca |
Molecular Weight (g/mol) | 199.89 |
MDL Number | MFCD00149608 |
InChI Key | WGEFECGEFUFIQW-UHFFFAOYSA-L |
IUPAC Name | calcium dibromide |
SMILES | [Ca++].[Br-].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15129323
|
Thermo Scientific Acros
450632500 |
250 g |
51.40 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15109323
|
Thermo Scientific Acros
450630025 |
2.5 kg |
292.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71626-99-8 | |
199.89 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
[Ca++].[Br-].[Br-] |
Br2Ca | |
MFCD00149608 | |
calcium dibromide |
Specifications
730.0°C | |
Br2Ca | |
MFCD00149608 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
calcium dibromide | |
199.9 | |
Plastic bottle |
71626-99-8 | |
CaBr2·xH2O | |
(10% in water) Clear colorless to slightly hazy | |
[Ca++].[Br-].[Br-] | |
199.89 | |
Extra Pure | |
Calcium bromide hydrate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only