missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium bromide, ultra dry , 99.978% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
CaBr2 is mainly used as a dense aqueous solution for drilling fluid. It is also used in neuroses medication, freezing mixtures, food preservatives, photography and fire retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
Boiling Point | 806°C to 812°C |
Melting Point | 730°C |
Color | White |
Physical Form | Beads |
Quantity | 5 g |
Linear Formula | CaBr2 |
Merck Index | 14,1654 |
Solubility Information | It is soluble in water,alcohol and acetone. |
Formula Weight | 199.9 |
Percent Purity | 99.978% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'