missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbide, 72-82%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 2300.0°C |
Melting Point | 2160.0°C |
Color | Gray |
Physical Form | Lump |
Quantity | 250 g |
Assay Percent Range | 12 to 23% (CaO) (Complexometry) |
Linear Formula | CaC2 |
Merck Index | 15, 1658 |
Solubility Information | Solubility in water: hydrolyses |
Formula Weight | 64.1 |
Show More |
Safety and Handling
Product Identifier
- Calcium carbide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P223-Do not allow contact with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Calcium carbide, Calcium oxide
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'